Learn More
Selleck Chemical LLC BI-3231-E1875-1G
SDP
Catalog No. 503137534
Encompass_Preferred
BI-3231 is the potent and selective chemical probe which inhibits hydroxysteroid 17 -dehydrogenase 13 (HSD17B13) with an IC50 of 1 nM and Ki of 0 7 nM in the enzymatic assay It has the potential to treat NASH and other liver diseases
Catalog No. 50-313-7534 Supplier Selleck Chemical LLC Supplier No. E18751G
BI-3231 is the potent and selective chemical probe which inhibits hydroxysteroid 17 -dehydrogenase 13 (HSD17B13) with an IC50 of 1 nM and Ki of 0 7 nM in the enzymatic assay It has the potential to treat NASH and other liver diseases