Learn More
Medchemexpress LLC BI-3406 | 2230836-55-0 | 99.95% | 462.46 | 10 MG
SDP
Catalog No. 5000365529
Encompass_Preferred
BI-3406 is an orally active, highly potent, and selective inhibitor of the interaction between KRAS and Son of Sevenless 1 (SOS1). It potently reduces the formation of GTP-loaded KRAS and inhibits MAPK pathway signaling, exhibiting anticancer activity. This compound shows activity on a broad panel of KRAS oncogenic variants, including all major G12 and G13 oncoproteins, and effectively limits cell proliferation in KRAS G12 or G13 mutant cells.
  • Highly potent and selective inhibitor of KRAS-SOS1 interaction
  • Reduces formation of GTP-loaded KRAS
  • Inhibits MAPK pathway signaling
  • Exhibits anticancer activity
  • Active against a broad panel of KRAS oncogenic variants (G12 and G13)
  • Limits cell proliferation in KRAS G12 or G13 mutant cells
  • Available for research use only
Catalog No. 50-003-65529 Supplier Medchemexpress LLC Supplier No. 10MGHY125817
BI-3406 is an orally active, highly potent, and selective inhibitor of the interaction between KRAS and Son of Sevenless 1 (SOS1). It potently reduces the formation of GTP-loaded KRAS and inhibits MAPK pathway signaling, exhibiting anticancer activity. This compound shows activity on a broad panel of KRAS oncogenic variants, including all major G12 and G13 oncoproteins, and effectively limits cell proliferation in KRAS G12 or G13 mutant cells.
  • Highly potent and selective inhibitor of KRAS-SOS1 interaction
  • Reduces formation of GTP-loaded KRAS
  • Inhibits MAPK pathway signaling
  • Exhibits anticancer activity
  • Active against a broad panel of KRAS oncogenic variants (G12 and G13)
  • Limits cell proliferation in KRAS G12 or G13 mutant cells
  • Available for research use only