Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bi-3406 | 2230836-55-0 | MFCD32197204 | 100.0% | 462.46 g/mol | C23H25F3N4O3 | 10MM 1ML
SDP

Catalog No. 5000238286
Encompass_Preferred

BI-3406 is a potent, selective, and orally bioavailable small-molecule inhibitor of the SOS1-KRAS protein-protein interaction. It is used as a research tool to reduce GTP-loaded RAS and to probe KRAS-dependent signaling in cellular and biochemical assays.

  • Potent SOS1-KRAS interaction inhibitor (IC50 ≈ 5-6 nM).
  • High selectivity for SOS1-mediated KRAS activation.
  • Reduces formation of GTP-loaded RAS in cellular models.
  • Available as a 10 mM stock solution in DMSO for in vitro use.
  • High purity suitable for research assays (supplier-reported ≈99.95%).
  • Molecular weight 462.46 g/mol and formula C23H25F3N4O3.
  • CAS number 2230836-55-0 for precise identification.

Catalog No. 50-002-38286 Supplier Medchemexpress LLC Supplier No. HY12581710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-3406 is a potent, selective, and orally bioavailable small-molecule inhibitor of the SOS1-KRAS protein-protein interaction. It is used as a research tool to reduce GTP-loaded RAS and to probe KRAS-dependent signaling in cellular and biochemical assays.

  • Potent SOS1-KRAS interaction inhibitor (IC50 ≈ 5-6 nM).
  • High selectivity for SOS1-mediated KRAS activation.
  • Reduces formation of GTP-loaded RAS in cellular models.
  • Available as a 10 mM stock solution in DMSO for in vitro use.
  • High purity suitable for research assays (supplier-reported ≈99.95%).
  • Molecular weight 462.46 g/mol and formula C23H25F3N4O3.
  • CAS number 2230836-55-0 for precise identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.