Learn More
TARGETMOL CHEMICALS INC BI-3406 5MG
SDP
Catalog No. 502581392
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-3406 is an orally active highly potent and selective between KRAS and Son of Sevenless 1 (SOS1) interaction inhibitor(IC50 6 nM)with anticancer activity. purity: 99%
Catalog No. 50-258-1392 Supplier TARGETMOL CHEMICALS INC Supplier No. T129795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-3406 is an orally active highly potent and selective between KRAS and Son of Sevenless 1 (SOS1) interaction inhibitor(IC50 6 nM)with anticancer activity. purity: 99%