Learn More
Medchemexpress LLC HY-125817 5mg Medchemexpress, BI-3406 CAS:2230836-55-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1258175MG
Encompass_Preferred
Medchemexpress, HY-125817 5mg BI-3406 CAS:2230836-55-0 BI-3406 (compound I-6) is a Son of Sevenless 1 (SOS1) inhibitor with an IC50 of 6 nM. BI-3406 has anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9696