Learn More
Medchemexpress LLC BI-3663 | 2341740-84-7 | 99.8% | 917.84 | 100 MG
SDP
Catalog No. 5000347083
Encompass_Preferred
BI-3663 is a highly selective PTK2/FAK PROTAC (DC50=30 nM) that utilizes Cereblon ligands to hijack E3 ligases for PTK2 degradation. It inhibits PTK2 with an IC50 of 18 nM. BI-3663 is composed of BI-4464 (HY-124625) linked to Pomalidomide (HY-10984) with a linker, and demonstrates anti-cancer activity.
  • Highly selective PTK2/FAK PROTAC
  • Inhibits PTK2 with an IC50 of 18 nM
  • Utilizes Cereblon ligands for PTK2 degradation
  • Composed of BI-4464 linked to Pomalidomide with a linker
  • Exhibits anti-cancer activity
  • Potently degrades PTK2 in Hep3B2.1-7 cells and A549 cells
Catalog No. 50-003-47083 Supplier Medchemexpress LLC Supplier No. HY11154650MG
BI-3663 is a highly selective PTK2/FAK PROTAC (DC50=30 nM) that utilizes Cereblon ligands to hijack E3 ligases for PTK2 degradation. It inhibits PTK2 with an IC50 of 18 nM. BI-3663 is composed of BI-4464 (HY-124625) linked to Pomalidomide (HY-10984) with a linker, and demonstrates anti-cancer activity.
  • Highly selective PTK2/FAK PROTAC
  • Inhibits PTK2 with an IC50 of 18 nM
  • Utilizes Cereblon ligands for PTK2 degradation
  • Composed of BI-4464 linked to Pomalidomide with a linker
  • Exhibits anti-cancer activity
  • Potently degrades PTK2 in Hep3B2.1-7 cells and A549 cells