Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-3663 | 2341740-84-7 | 99.8% | 917.84 | 25 MG
SDP

Catalog No. 5000346773
Encompass_Preferred

BI-3663 is a highly selective PTK2/FAK PROTAC that uses Cereblon ligands to hijack E3 ligases for PTK2 degradation. It inhibits PTK2 with an IC50 of 18 nM. BI-3663 is composed of BI-4464 linked to Pomalidomide with a linker, and demonstrates anti-cancer activity. It potently degrades PTK2 in Hep3B2.1-7 cells and A549 cells.

  • Highly selective PTK2/FAK PROTAC (DC50=30 nM)
  • Inhibits PTK2 with an IC50 of 18 nM
  • Exhibits anti-cancer activity
  • Potently degrades PTK2 in Hep3B2.1-7 cells and A549 cells (pDC50s of 7.6 and 7.9, respectively)
  • For research use only

Catalog No. 50-003-46773 Supplier Medchemexpress LLC Supplier No. 25MGHY111546
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-3663 is a highly selective PTK2/FAK PROTAC that uses Cereblon ligands to hijack E3 ligases for PTK2 degradation. It inhibits PTK2 with an IC50 of 18 nM. BI-3663 is composed of BI-4464 linked to Pomalidomide with a linker, and demonstrates anti-cancer activity. It potently degrades PTK2 in Hep3B2.1-7 cells and A549 cells.

  • Highly selective PTK2/FAK PROTAC (DC50=30 nM)
  • Inhibits PTK2 with an IC50 of 18 nM
  • Exhibits anti-cancer activity
  • Potently degrades PTK2 in Hep3B2.1-7 cells and A549 cells (pDC50s of 7.6 and 7.9, respectively)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.