Learn More
Medchemexpress LLC BI-3663 | 2341740-84-7 | 99.84% | 917.84 | 1 ML
SDP
Catalog No. 5000347164
Encompass_Preferred
BI-3663 is a highly selective PTK2/FAK PROTAC that hijacks E3 ligases for PTK2 degradation using Cereblon ligands. It inhibits PTK2 with an IC50 of 18 nM. This PROTAC is formed by linking BI-4464 to Pomalidomide. It exhibits anti-cancer activity and potently degrades PTK2 in Hep3B2.1-7 and A549 cells.
  • Highly selective PTK2/FAK PROTAC.
  • Inhibits PTK2 with an IC50 of 18 nM.
  • Utilizes Cereblon ligands for E3 ligase hijacking and PTK2 degradation.
  • Exhibits anti-cancer activity.
  • Potently degrades PTK2 in Hep3B2.1-7 and A549 cells.
Catalog No. 50-003-47164 Supplier Medchemexpress LLC Supplier No. HY111546100MG
BI-3663 is a highly selective PTK2/FAK PROTAC that hijacks E3 ligases for PTK2 degradation using Cereblon ligands. It inhibits PTK2 with an IC50 of 18 nM. This PROTAC is formed by linking BI-4464 to Pomalidomide. It exhibits anti-cancer activity and potently degrades PTK2 in Hep3B2.1-7 and A549 cells.
  • Highly selective PTK2/FAK PROTAC.
  • Inhibits PTK2 with an IC50 of 18 nM.
  • Utilizes Cereblon ligands for E3 ligase hijacking and PTK2 degradation.
  • Exhibits anti-cancer activity.
  • Potently degrades PTK2 in Hep3B2.1-7 and A549 cells.