Learn More
Medchemexpress LLC HY-111546 5mg Medchemexpress, BI-3663 CAS:2341740-84-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115465MG
Encompass_Preferred
Medchemexpress, HY-111546 5mg BI-3663 CAS:2341740-84-7 BI-3663 is a highly selective PTK2/FAK PROTAC, with cereblon ligands to hijack E3 ligases for PTK2 degradation. BI-3663 inhibits PTK2 with an IC50 of 18 nM. BI-3663 is a PROTAC that composes of BI-4464 (HY-124625) linked to Pomalidomide (HY-10984) with a linker. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8911