Learn More
Selleck Chemical LLC BI-3802 5mg
SDP
Supplier:  Selleck Chemical LLC S69375MG
Encompass_Preferred
BI-3802 is a highly potent oncogenic transcription factor BCL6 degrader with strong de-repression of target genes anti-proliferative effects. BI-3802 inhibits the BTB domain of BCL6 with IC50 of ?3 nM. BI-3802 exhibits antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3239