Learn More
Selleck Chemical LLC BI-3802
SDP
Catalog No. 5000002239
Encompass_Preferred
BI-3802 is a highly potent oncogenic transcription factor BCL6 degrader with strong de-repression of target genes anti-proliferative effects BI-3802 inhibits the BTB domain of BCL6 with IC50 of 3 nM BI-3802 exhibits antitumor activity
Catalog No. 50-000-02239 Supplier Selleck Chemical LLC Supplier No. S693725MG
BI-3802 is a highly potent oncogenic transcription factor BCL6 degrader with strong de-repression of target genes anti-proliferative effects BI-3802 inhibits the BTB domain of BCL6 with IC50 of 3 nM BI-3802 exhibits antitumor activity