Learn More
Medchemexpress LLC HY-111381 10mg Medchemexpress, BI-3812 CAS:2166387-64-8 Purity:>98%
SDP
Catalog No. 501968846
Encompass_Preferred
Medchemexpress, HY-111381 10mg BI-3812 CAS:2166387-64-8 BI-3812 is potent and efficacious BCL6 inhibitor, inhibiting the BTB domain of BCL6, with an IC50 of ≤3 nM; BI-3812 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8846 Supplier Medchemexpress LLC Supplier No. HY11138110MG
Add to Cart
Add to Cart
Medchemexpress, HY-111381 10mg BI-3812 CAS:2166387-64-8 BI-3812 is potent and efficacious BCL6 inhibitor, inhibiting the BTB domain of BCL6, with an IC50 of ≤3 nM; BI-3812 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.