Learn More
Medchemexpress LLC Bi-3812 | 2166387-64-8 | 99.3% | 558.03 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1113811MG
Encompass_Preferred
BI-3812 is identified as a chemical probe and a highly efficient BCL6 inhibitor that suppresses the BTB domain of BCL6 with an IC50 of ≤ 3 nM, demonstrating antitumor activity.
  • Highly efficient BCL6 inhibitor.
  • Capable of suppressing the BTB domain of BCL6.
  • Exhibits antitumor activity.
  • IC50 value of ≤ 3 nM for BCL6 BTB.
  • IC50 of 40 nM for cellular BCL6.
Catalog No. 50-003-43125