Learn More
Medchemexpress LLC BI-3812 | 2166387-64-8 | 99.3% | 558.03 | 100 MG
Supplier: Medchemexpress LLC HY111381100MG
BI-3812 is a chemical probe and a highly efficient BCL6 inhibitor. It suppresses the BTB domain of BCL6 with an IC50 value of ≤ 3 nM, demonstrating antitumor activity. This product is intended for research use only and not for patient use.
- Highly efficient BCL6 inhibitor
- Suppresses the BTB domain of BCL6 with an IC50 value of ≤ 3 nM
- Exhibits antitumor activity
- Cellular BCL6 IC50 of 40 nM
- Available as a chemical probe