Learn More
Medchemexpress LLC BI-3812 | 2166387-64-8 | 99.3% | 558.03 | 50 MG
SDP
Catalog No. 5000343795
Encompass_Preferred
BI-3812 is a chemical probe and a highly efficient BCL6 inhibitor capable of suppressing the BTB domain of BCL6. It has an IC50 value of ≤ 3 nM and exhibits antitumor activity. This product is for research use only and not sold to patients.
  • Highly efficient BCL6 inhibitor
  • Suppresses the BTB domain of BCL6
  • IC50 value of ≤ 3 nM
  • Exhibits antitumor activity
Catalog No. 50-003-43795 Supplier Medchemexpress LLC Supplier No. HY11138150MG
Add to Cart
Add to Cart
BI-3812 is a chemical probe and a highly efficient BCL6 inhibitor capable of suppressing the BTB domain of BCL6. It has an IC50 value of ≤ 3 nM and exhibits antitumor activity. This product is for research use only and not sold to patients.
  • Highly efficient BCL6 inhibitor
  • Suppresses the BTB domain of BCL6
  • IC50 value of ≤ 3 nM
  • Exhibits antitumor activity