Learn More
TARGETMOL CHEMICALS INC BI-3812 5MG
SDP
Catalog No. 502581281
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-3812 is an inhibitor of BCL6. Which inhibiting the BTB domain of BCL6 (IC50 3 nM). BI-3812 also has antitumor activity. purity: 99%
Catalog No. 50-258-1281 Supplier TARGETMOL CHEMICALS INC Supplier No. T145615MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-3812 is an inhibitor of BCL6. Which inhibiting the BTB domain of BCL6 (IC50 3 nM). BI-3812 also has antitumor activity. purity: 99%