Learn More
Selleck Chemical LLC BI-3812 S8735-5MG
SDP
Catalog No. 5000024241
Encompass_Preferred
BI-3812 is a highly potent and efficacious BCL6 inhibitor probe compound with an IC50 less than 3 nM
Catalog No. 50-000-24241 Supplier Selleck Chemical LLC Supplier No. S87355MG
BI-3812 is a highly potent and efficacious BCL6 inhibitor probe compound with an IC50 less than 3 nM