Learn More
Medchemexpress LLC BI-4020 | 2664214-60-0 | 99.3% | 10 MG
SDP
Catalog No. 5000340700
Encompass_Preferred
BI-4020 is a fourth-generation, orally active, and non-covalent EGFR tyrosine kinase inhibitor. It inhibits the triple mutant EGFR del19 T790M C797S variant (IC50=0.2 nM in BaF3 cell lines), the double mutant EGFR del19 T790M, and primary mutant EGFR del19 (IC50=1 nM). It also shows activity against EGFR wt (IC50=190 nM). This product demonstrates high kinome selectivity and good DMPK properties and is for research use only.
  • Fourth-generation, orally active, and non-covalent EGFR tyrosine kinase inhibitor
  • Inhibits triple mutant EGFR del19 T790M C797S variant (IC50=0.2 nM)
  • Inhibits double mutant EGFR del19 T790M and primary mutant EGFR del19 (IC50=1 nM)
  • Shows activity against EGFR wt (IC50=190 nM)
  • Demonstrates high kinome selectivity and good DMPK properties
  • Leads to tumor regressions in human PC-9 triple mutant NSCLC xenograft model in mice
Catalog No. 50-003-40700 Supplier Medchemexpress LLC Supplier No. HY12955010MG
BI-4020 is a fourth-generation, orally active, and non-covalent EGFR tyrosine kinase inhibitor. It inhibits the triple mutant EGFR del19 T790M C797S variant (IC50=0.2 nM in BaF3 cell lines), the double mutant EGFR del19 T790M, and primary mutant EGFR del19 (IC50=1 nM). It also shows activity against EGFR wt (IC50=190 nM). This product demonstrates high kinome selectivity and good DMPK properties and is for research use only.
  • Fourth-generation, orally active, and non-covalent EGFR tyrosine kinase inhibitor
  • Inhibits triple mutant EGFR del19 T790M C797S variant (IC50=0.2 nM)
  • Inhibits double mutant EGFR del19 T790M and primary mutant EGFR del19 (IC50=1 nM)
  • Shows activity against EGFR wt (IC50=190 nM)
  • Demonstrates high kinome selectivity and good DMPK properties
  • Leads to tumor regressions in human PC-9 triple mutant NSCLC xenograft model in mice