Learn More
TARGETMOL CHEMICALS INC BI-4020 5MG
SDP
Catalog No. 502581711
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-4020 is a fourth-generation orally active and non-covalent inhibitor of EGFR tyrosine kinase. BI-4020 exhibits activity agains the triple mutant EGFR del19 T790M C797S variant (IC50=0.2 nM in BaF3 cell lines) the double mutant EGFR del19 T790M and primary mutant EGFR del19 (IC50=1 nM) and EGFR wt (IC50=190 nM). BI-4020 exhibits high kinome selectivity and good DMPK properties. purity: 98%
Catalog No. 50-258-1711 Supplier TARGETMOL CHEMICALS INC Supplier No. T105345MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-4020 is a fourth-generation orally active and non-covalent inhibitor of EGFR tyrosine kinase. BI-4020 exhibits activity agains the triple mutant EGFR del19 T790M C797S variant (IC50=0.2 nM in BaF3 cell lines) the double mutant EGFR del19 T790M and primary mutant EGFR del19 (IC50=1 nM) and EGFR wt (IC50=190 nM). BI-4020 exhibits high kinome selectivity and good DMPK properties. purity: 98%