Learn More
Medchemexpress LLC HY-112831 10mg Medchemexpress, BI-409306 CAS:1189767-28-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11283110MG
Encompass_Preferred
Medchemexpress, HY-112831 10mg BI-409306 CAS:1189767-28-9 BI-409306 is a potent and selective PDE9A inhibitor, with an IC50 of 52 nM, and shows weak activity against other PDEs, such as PDE1A (IC50, 1.4 µM), PDE1C (IC50, 1.0 µM), PDE2A, PDE3A, PDE4B, PDE5A, PDE6AB, PDE7A, and PDE10A (IC50 all > 10 μM); BI-409306 can be used in the research of memory enhancement in CNS disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9187