Learn More
Medchemexpress LLC BI-4142 | 2682003-36-5 | 96.5% | 521.57 | 100 MG
SDP
Catalog No. 5000361232
Encompass_Preferred
BI-4142 is a potent, highly selective, and orally active HER2 inhibitor with an IC50 of 5 nM. It demonstrates significant antiproliferative activity against various tumor cells and inhibits HER2-dependent cell lines and downstream signaling. In vivo studies have shown that BI-4142 effectively inhibits tumor growth and oncogenic signaling in xenograft models with good oral bioavailability.
  • Potent, highly selective, and orally active HER2 inhibitor.
  • Inhibits HER2-dependent cell lines and downstream signaling.
  • Demonstrates antiproliferative activity against tumor cells in vitro.
  • Effectively inhibits tumor growth and oncogenic signaling in vivo.
  • Good oral bioavailability in pharmacokinetic studies.
  • Soluble in DMSO for in vitro applications.
Catalog No. 50-003-61232 Supplier Medchemexpress LLC Supplier No. HY150024100MG
BI-4142 is a potent, highly selective, and orally active HER2 inhibitor with an IC50 of 5 nM. It demonstrates significant antiproliferative activity against various tumor cells and inhibits HER2-dependent cell lines and downstream signaling. In vivo studies have shown that BI-4142 effectively inhibits tumor growth and oncogenic signaling in xenograft models with good oral bioavailability.
  • Potent, highly selective, and orally active HER2 inhibitor.
  • Inhibits HER2-dependent cell lines and downstream signaling.
  • Demonstrates antiproliferative activity against tumor cells in vitro.
  • Effectively inhibits tumor growth and oncogenic signaling in vivo.
  • Good oral bioavailability in pharmacokinetic studies.
  • Soluble in DMSO for in vitro applications.