Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-4142 | 2682003-36-5 | 96.5% | 521.57 | 25 MG
SDP

Catalog No. 5000360391
Encompass_Preferred

BI-4142 is a potent, highly selective, and orally active HER2 inhibitor with an IC50 of 5 nM. It demonstrates antiproliferative activity against tumor cells and inhibits tumor growth and oncogenic signaling in animal models. The compound exhibits good permeability and no PgP-mediated efflux liability in the CaCo-2 assay.

  • Potent, highly selective, and orally active HER2 inhibitor
  • IC50 of 5 nM against HER2
  • Inhibits tumor growth and oncogenic signaling
  • Displays good permeability and no PgP-mediated efflux liability
  • Inhibits HER2-dependent cell lines and downstream signaling
  • Target: EGFR, JAK/STAT Signaling, Protein Tyrosine Kinase/RTK
  • Soluble in DMSO at 100 mg/mL (191.73 mM)

Catalog No. 50-003-60391 Supplier Medchemexpress LLC Supplier No. 25MGHY150024
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-4142 is a potent, highly selective, and orally active HER2 inhibitor with an IC50 of 5 nM. It demonstrates antiproliferative activity against tumor cells and inhibits tumor growth and oncogenic signaling in animal models. The compound exhibits good permeability and no PgP-mediated efflux liability in the CaCo-2 assay.

  • Potent, highly selective, and orally active HER2 inhibitor
  • IC50 of 5 nM against HER2
  • Inhibits tumor growth and oncogenic signaling
  • Displays good permeability and no PgP-mediated efflux liability
  • Inhibits HER2-dependent cell lines and downstream signaling
  • Target: EGFR, JAK/STAT Signaling, Protein Tyrosine Kinase/RTK
  • Soluble in DMSO at 100 mg/mL (191.73 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.