Learn More
Medchemexpress LLC Bi-4394 (MMP13-IN-3) | 1222173-37-6 | 99.1% | 446.46 | C24H22N4O5 | 25MG
SDP
Catalog No. 5000235071
Encompass_Preferred
BI-4394 is a research-grade small molecule inhibitor of matrix metalloproteinase-13 (MMP-13). It is described as a potent, selective, and orally active MMP-13 inhibitor (IC50 = 1 nM) with greater than 1,000-fold selectivity over other MMP isoforms; supplied for biochemical and preclinical research as a solid or as a 10 mM solution in DMSO.
  • Potent MMP-13 inhibition (IC50 = 1 nM).
  • Greater than 1,000-fold selectivity over other MMP isoforms.
  • High purity (99.09%).
  • Chemical formula C24H22N4O5; molecular weight 446.46.
  • Available in multiple pack sizes, including 25 mg, and as a 10 mM DMSO solution.
  • Suitable for biochemical and preclinical research applications.
Catalog No. 50-002-35071 Supplier Medchemexpress LLC Supplier No. HY12402925MG
BI-4394 is a research-grade small molecule inhibitor of matrix metalloproteinase-13 (MMP-13). It is described as a potent, selective, and orally active MMP-13 inhibitor (IC50 = 1 nM) with greater than 1,000-fold selectivity over other MMP isoforms; supplied for biochemical and preclinical research as a solid or as a 10 mM solution in DMSO.
  • Potent MMP-13 inhibition (IC50 = 1 nM).
  • Greater than 1,000-fold selectivity over other MMP isoforms.
  • High purity (99.09%).
  • Chemical formula C24H22N4O5; molecular weight 446.46.
  • Available in multiple pack sizes, including 25 mg, and as a 10 mM DMSO solution.
  • Suitable for biochemical and preclinical research applications.