Learn More
Selleck Chemical LLC BI-4464
SDP
Catalog No. 5000002575
Encompass_Preferred
BI-4464 is a highly selective ATP competitive inhibitor of PTK2/FAK with an IC50 of 17 nM
Catalog No. 50-000-02575 Supplier Selleck Chemical LLC Supplier No. S65455MG
BI-4464 is a highly selective ATP competitive inhibitor of PTK2/FAK with an IC50 of 17 nM