Learn More
Medchemexpress LLC BI-4464 | 1227948-02-8 | 10MG
SDP
Catalog No. 5000235516
Encompass_Preferred
BI-4464 is a highly selective ATP competitive PTK2/FAK protein kinase inhibitor with an IC50 value of 17 nM BI-4464 can be used as a PTK2 ligand for constructing proteolysis targeting chimeras (PROTAC)
Catalog No. 50-002-35516 Supplier Medchemexpress LLC Supplier No. HY12462510MG
BI-4464 is a highly selective ATP competitive PTK2/FAK protein kinase inhibitor with an IC50 value of 17 nM BI-4464 can be used as a PTK2 ligand for constructing proteolysis targeting chimeras (PROTAC)