Learn More
Medchemexpress LLC BI-4464 2 | 1227948-02-8 | 98.6% | 555.55 Da | C28H28F3N5O4 | 25MG
SDP
Catalog No. 5000239377
Encompass_Preferred
BI-4464 is a selective ATP-competitive inhibitor of PTK2/FAK used as a research reagent for biochemical and cellular studies. It is provided with a reported purity of 98.64%, molecular formula C28H28F3N5O4, and a molecular weight of 555.55 Da. Intended for assay development, kinase profiling, and use as a ligand in targeted degradation studies; not for human or clinical use.
  • High reported purity suitable for research applications.
  • Selective ATP-competitive PTK2/FAK inhibition profile.
  • Applicable to assay development and kinase profiling.
  • Useful as a ligand in PROTAC and targeted degradation research.
  • Available in multiple mg-scale quantities for laboratory use.
  • Research use only; not intended for human or clinical use.
Catalog No. 50-002-39377 Supplier Medchemexpress LLC Supplier No. HY12462525MG
BI-4464 is a selective ATP-competitive inhibitor of PTK2/FAK used as a research reagent for biochemical and cellular studies. It is provided with a reported purity of 98.64%, molecular formula C28H28F3N5O4, and a molecular weight of 555.55 Da. Intended for assay development, kinase profiling, and use as a ligand in targeted degradation studies; not for human or clinical use.
  • High reported purity suitable for research applications.
  • Selective ATP-competitive PTK2/FAK inhibition profile.
  • Applicable to assay development and kinase profiling.
  • Useful as a ligand in PROTAC and targeted degradation research.
  • Available in multiple mg-scale quantities for laboratory use.
  • Research use only; not intended for human or clinical use.