Learn More
Medchemexpress LLC HY-124625 5mg Medchemexpress, BI-4464 CAS:1227948-02-8 Purity:>98%
SDP
Catalog No. 501932594
Encompass_Preferred
Medchemexpress, HY-124625 5mg BI-4464 CAS:1227948-02-8 BI-4464 is a highly selective ATP competitive inhibitor of PTK2/FAK , with an IC 50 of 17 nM. A PTK2 ligand for PROTAC [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2594 Supplier Medchemexpress LLC Supplier No. HY1246255MG
Medchemexpress, HY-124625 5mg BI-4464 CAS:1227948-02-8 BI-4464 is a highly selective ATP competitive inhibitor of PTK2/FAK , with an IC 50 of 17 nM. A PTK2 ligand for PROTAC [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.