Learn More
Medchemexpress LLC BI-4732 | 2769715-68-4 | 99.3% | 592.69 | 50 MG
SDP
Catalog No. 5000391773
Encompass_Preferred
BI-4732 is an orally active, reversible, and ATP-competitive EGFR inhibitor that can cross the blood-brain barrier. It selectively inhibits the kinase activity of EGFR L858R, T790M, and C797S with high potency, while sparing wild-type EGFR. BI-4732 also reduces the phosphorylation of AKT, ERK, and S6K, and has shown excellent intracranial anti-tumor efficacy in a YU-1097 xenograft model. This product is suitable for non-small cell lung cancer (NSCLC) research.
  • Orally active, reversible, and ATP-competitive EGFR inhibitor
  • Penetrates the blood-brain barrier
  • Selectively inhibits EGFR L858R, T790M, and C797S with high potency
  • Reduces phosphorylation of AKT, ERK, and S6K
  • Demonstrates excellent intracranial anti-tumor efficacy in xenograft models
  • Suitable for non-small cell lung cancer (NSCLC) studies
Catalog No. 50-003-91773 Supplier Medchemexpress LLC Supplier No. HY16127550MG
BI-4732 is an orally active, reversible, and ATP-competitive EGFR inhibitor that can cross the blood-brain barrier. It selectively inhibits the kinase activity of EGFR L858R, T790M, and C797S with high potency, while sparing wild-type EGFR. BI-4732 also reduces the phosphorylation of AKT, ERK, and S6K, and has shown excellent intracranial anti-tumor efficacy in a YU-1097 xenograft model. This product is suitable for non-small cell lung cancer (NSCLC) research.
  • Orally active, reversible, and ATP-competitive EGFR inhibitor
  • Penetrates the blood-brain barrier
  • Selectively inhibits EGFR L858R, T790M, and C797S with high potency
  • Reduces phosphorylation of AKT, ERK, and S6K
  • Demonstrates excellent intracranial anti-tumor efficacy in xenograft models
  • Suitable for non-small cell lung cancer (NSCLC) studies