Learn More
Medchemexpress LLC HY-126253 5mg Medchemexpress, BI-4916 CAS:2244451-48-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1262535MG
Encompass_Preferred
Medchemexpress, HY-126253 5mg BI-4916 CAS:2244451-48-5 BI-4916 is a prodrug of BI-4924. BI-4924 is a NADH/NAD+-competitive PHGDH inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0048