Learn More
TARGETMOL CHEMICALS INC BI-6015 25MG
SDP
Catalog No. 502580993
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI 6015 is an antagonist of HNF4(alpha). HNF4(alpha) is a nuclear receptor that regulates gene expression in the intestine cells liver cells and pancreas-(beta) cells. purity: 99%
Catalog No. 50-258-0993 Supplier TARGETMOL CHEMICALS INC Supplier No. T2186725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI 6015 is an antagonist of HNF4(alpha). HNF4(alpha) is a nuclear receptor that regulates gene expression in the intestine cells liver cells and pancreas-(beta) cells. purity: 99%