Learn More
Medchemexpress LLC Bi 639667 (compound 19n) | 1295298-26-8 | 98.9% | 10 MG
SDP
Catalog No. 5000233522
Encompass_Preferred
BI 639667 is a small-molecule CCR1 antagonist chemical probe with low-nanomolar potency in biochemical and cellular assays. It is intended for in vitro pharmacology and target-validation studies exploring CCR1-mediated chemotaxis and signaling.
  • Chemical probe for CCR1 with potent receptor antagonism.
  • Ca2+ flux IC50 ≈ 1.8 nM; SPA binding ~5.4 nM; chemotaxis IC50 ~2.4 nM.
  • CAS 1295298-26-8; molecular formula C22H18FN5O3S; molecular weight 451.47 Da.
  • High chemical purity (reported 98.87%).
  • Supplied as a 10 mg sample suitable for in vitro assays.
Catalog No. 50-002-33522 Supplier Medchemexpress LLC Supplier No. HY12058810MG
BI 639667 is a small-molecule CCR1 antagonist chemical probe with low-nanomolar potency in biochemical and cellular assays. It is intended for in vitro pharmacology and target-validation studies exploring CCR1-mediated chemotaxis and signaling.
  • Chemical probe for CCR1 with potent receptor antagonism.
  • Ca2+ flux IC50 ≈ 1.8 nM; SPA binding ~5.4 nM; chemotaxis IC50 ~2.4 nM.
  • CAS 1295298-26-8; molecular formula C22H18FN5O3S; molecular weight 451.47 Da.
  • High chemical purity (reported 98.87%).
  • Supplied as a 10 mg sample suitable for in vitro assays.