Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI 653048 | 1198784-72-3 | 99.8% | 25 MG
SDP

Catalog No. 5000342226
Encompass_Preferred

BI 653048 is a chemical probe, a selective and orally active nonsteroidal glucocorticoid (GC) agonist with an IC50 value of 55 nM. It also acts as an HCV NS3 protease inhibitor.

  • Inhibits CP1A2, CYP2D6, CYP2C9, CYP2C19, and CYP3A4 isoforms' activity.
  • Reduces affinity for the hERG ion channel (IC50 > 30 μM).
  • Can reduce viral loads infected with the hepatitis C virus.

Catalog No. 50-003-42226 Supplier Medchemexpress LLC Supplier No. 25MGHY12946
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI 653048 is a chemical probe, a selective and orally active nonsteroidal glucocorticoid (GC) agonist with an IC50 value of 55 nM. It also acts as an HCV NS3 protease inhibitor.

  • Inhibits CP1A2, CYP2D6, CYP2C9, CYP2C19, and CYP3A4 isoforms' activity.
  • Reduces affinity for the hERG ion channel (IC50 > 30 μM).
  • Can reduce viral loads infected with the hepatitis C virus.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.