Learn More
Medchemexpress LLC BI-671800 | 1093108-50-9 | 99.4% | 501.50 | 1 ML
SDP
Catalog No. 5000436844
Encompass_Preferred
BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. It has potential for the treatment of poorly controlled asthma.
  • Hcrth2: 4.5 nM (IC50)
  • Mcrth2: 3.7 μM (IC50)
  • Exhibits low nM potency as an antagonist of human or mouse CRTH2 in transfected cells.
  • Shows significant inhibition of AHR in mice at 0.1-10 mg/kg, i.g.
  • Effectively blocks edema formation and greatly reduces inflammatory infiltrate and skin pathology observed in drug vehicle-treated animals.
Catalog No. 50-004-36844 Supplier Medchemexpress LLC Supplier No. HY11414110MM/1ML
BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. It has potential for the treatment of poorly controlled asthma.
  • Hcrth2: 4.5 nM (IC50)
  • Mcrth2: 3.7 μM (IC50)
  • Exhibits low nM potency as an antagonist of human or mouse CRTH2 in transfected cells.
  • Shows significant inhibition of AHR in mice at 0.1-10 mg/kg, i.g.
  • Effectively blocks edema formation and greatly reduces inflammatory infiltrate and skin pathology observed in drug vehicle-treated animals.