Learn More
Medchemexpress LLC HY-114141 10mg Medchemexpress, BI-671800 CAS:1093108-50-9 Purity:>98%
SDP
Catalog No. 501969287
Encompass_Preferred
Medchemexpress, HY-114141 10mg BI-671800 CAS:1093108-50-9 BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. BI-671800 has potential for the treatment of poorly controlled asthma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9287 Supplier Medchemexpress LLC Supplier No. HY11414110MG
Medchemexpress, HY-114141 10mg BI-671800 CAS:1093108-50-9 BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. BI-671800 has potential for the treatment of poorly controlled asthma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.