Learn More
Medchemexpress LLC BI-6901 10mM | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY11683510MM 1ML
Encompass_Preferred
BI 6901 is a potent selective CCR10 antagonist (pIC50 9 0) BI 6901 shows high selectivity over other GPCRs including a number of other chemokine receptors BI 6901 is efficacious in the murine DNFB model of contact hypersensitivity and can be used for inflammation research[1]
Catalog No. 50-002-27659