Learn More
TARGETMOL CHEMICALS INC BI-69A11 25MG
SDP
Catalog No. 502877529
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BI-69A11 ((E)-3-(1H-Benzo[d]imidazol-2-yl)-1-(6-chloro-2-hydroxy-4-phenylquinolin-3-yl)prop-2-en-1-one) is a dual AKT and NFkB pathway inhibitor. It enhances the susceptibility of colon cancer cells to mda-7/IL-24-induced growth inhibition by targeting Akt. Purity 96.97%
Catalog No. 50-287-7529 Supplier TARGETMOL CHEMICALS INC Supplier No. T2515025MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BI-69A11 ((E)-3-(1H-Benzo[d]imidazol-2-yl)-1-(6-chloro-2-hydroxy-4-phenylquinolin-3-yl)prop-2-en-1-one) is a dual AKT and NFkB pathway inhibitor. It enhances the susceptibility of colon cancer cells to mda-7/IL-24-induced growth inhibition by targeting Akt. Purity 96.97%