Learn More
TARGETMOL CHEMICALS INC BI-6C9 5MG
SDP
Catalog No. 502579728
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-6C9 is an inhibitor of tBid (Kd = 20 uM).BI-6c9 prevented MOMP and mitochondrial fission and protected the cells from cell death. purity: 98%
Catalog No. 50-257-9728 Supplier TARGETMOL CHEMICALS INC Supplier No. T58275MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-6C9 is an inhibitor of tBid (Kd = 20 uM).BI-6c9 prevented MOMP and mitochondrial fission and protected the cells from cell death. purity: 98%