Learn More
Medchemexpress LLC HY-103661 5mg Medchemexpress, BI-6C9 CAS:791835-21-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1036615MG
Encompass_Preferred
Medchemexpress, HY-103661 5mg BI-6C9 CAS:791835-21-7 BI-6C9 is a highly specific BH3 interacting domain (Bid) inhibitor, which prevents mitochondrial outer membrane potential (MOMP) and mitochondrial fission, and protects the cells from mitochondrial apoptosis inducing factor (AIF) release and caspase-independent cell death in neurons. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8153