Learn More
Medchemexpress LLC Bi-6c9 | 791835-21-7 | MFCD07784498 | 98.5% | 471.59 | C23H25N3O4S2 | 25 MG
SDP
Catalog No. 5000246860
Encompass_Preferred
BI-6C9 is a small-molecule inhibitor of the BH3 interacting domain (Bid) used in research to modulate mitochondrial outer membrane permeabilization and mitochondrial fission. The compound is supplied as a characterized solid reagent with documented solubility and storage recommendations for laboratory use.
  • Acts as a Bid inhibitor to modulate MOMP and mitochondrial fission.
  • High purity (≈98.5%) for research applications.
  • Well characterized physicochemical data: molecular weight 471.59, formula C23H25N3O4S2.
  • Soluble in DMSO; documented in vitro and in vivo formulation protocols.
  • Available in small solid quantities including 25 mg for laboratory experiments.
  • Powder stable when stored at -20°C; solution stability notes available.
Catalog No. 50-002-46860 Supplier Medchemexpress LLC Supplier No. HY10366125MG
BI-6C9 is a small-molecule inhibitor of the BH3 interacting domain (Bid) used in research to modulate mitochondrial outer membrane permeabilization and mitochondrial fission. The compound is supplied as a characterized solid reagent with documented solubility and storage recommendations for laboratory use.
  • Acts as a Bid inhibitor to modulate MOMP and mitochondrial fission.
  • High purity (≈98.5%) for research applications.
  • Well characterized physicochemical data: molecular weight 471.59, formula C23H25N3O4S2.
  • Soluble in DMSO; documented in vitro and in vivo formulation protocols.
  • Available in small solid quantities including 25 mg for laboratory experiments.
  • Powder stable when stored at -20°C; solution stability notes available.