Learn More
Selleck Chemical LLC BI-6C9-E0808-25MG
SDP
Catalog No. 503137154
Encompass_Preferred
BI-6C9 is a highly specific BH3 interacting domain (Bid) inhibitor which prevents mitochondrial outer membrane potential (MOMP) and mitochondrial fission and protects the cells from mitochondrial apoptosis inducing factor (AIF) release and caspase-independent cell death in neurons
Catalog No. 50-313-7154 Supplier Selleck Chemical LLC Supplier No. E080825MG
BI-6C9 is a highly specific BH3 interacting domain (Bid) inhibitor which prevents mitochondrial outer membrane potential (MOMP) and mitochondrial fission and protects the cells from mitochondrial apoptosis inducing factor (AIF) release and caspase-independent cell death in neurons