Learn More
Medchemexpress LLC BI-7273 | 1883429-21-7 | 99.8% | 353.41 | 5 MG
SDP
Catalog No. 5000364561
Encompass_Preferred
BI-7273 is a selective and cell-permeable BRD9 inhibitor, exhibiting an IC50 of 19 nM and a Kd of 0.75 nM. It also demonstrates significant activity against BRD7, with an IC50 of 117 nM and a Kd of 0.3 nM. Originally designed by Boehringer Ingelheim, this compound is suitable for various laboratory applications.
  • Selective BRD9 inhibitor
  • Cell-permeable compound
  • High effect on BRD7
  • Active in U2OS cell lines at 1 μM
  • Blocks EOL-1 cell proliferation
Catalog No. 50-003-64561 Supplier Medchemexpress LLC Supplier No. 5MGHY100351
BI-7273 is a selective and cell-permeable BRD9 inhibitor, exhibiting an IC50 of 19 nM and a Kd of 0.75 nM. It also demonstrates significant activity against BRD7, with an IC50 of 117 nM and a Kd of 0.3 nM. Originally designed by Boehringer Ingelheim, this compound is suitable for various laboratory applications.
  • Selective BRD9 inhibitor
  • Cell-permeable compound
  • High effect on BRD7
  • Active in U2OS cell lines at 1 μM
  • Blocks EOL-1 cell proliferation