Learn More
Selleck Chemical LLC BI-7273
SDP
Catalog No. 5000000898
Encompass_Preferred
BI-7273 is a potent selective and cell-permeable BRD9 BD inhibitor with IC50s of 19 nM and 117 nM for BRD9 and BRD7 respectively in alpha assay
Catalog No. 50-000-00898 Supplier Selleck Chemical LLC Supplier No. S817925MG
BI-7273 is a potent selective and cell-permeable BRD9 BD inhibitor with IC50s of 19 nM and 117 nM for BRD9 and BRD7 respectively in alpha assay