Learn More
Selleck Chemical LLC BI-7273 5mg 1883429-21-7
SDP
Supplier:  Selleck Chemical LLC S81795MG
Encompass_Preferred
BI-7273 is a potent, selective, and cell-permeable BRD9 BD inhibitor with IC50s of 19 nM and 117 nM for BRD9 and BRD7 respectively in alpha assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5753