Learn More
Medchemexpress LLC HY-111925 10mg Medchemexpress, BI-749327 CAS:2361241-23-6 Purity:>98%
SDP
Catalog No. 501968985
Encompass_Preferred
Medchemexpress, HY-111925 10mg BI-749327 CAS:2361241-23-6 BI-749327 is a potent, high selectivity and orally bioavailable TRPC6 antagonist, with IC50s of 13 nM, 19 nM and 15 nM for mouse, human and guinea pig TRPC6, respectively. BI-749327 is 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8985 Supplier Medchemexpress LLC Supplier No. HY11192510MG
Medchemexpress, HY-111925 10mg BI-749327 CAS:2361241-23-6 BI-749327 is a potent, high selectivity and orally bioavailable TRPC6 antagonist, with IC50s of 13 nM, 19 nM and 15 nM for mouse, human and guinea pig TRPC6, respectively. BI-749327 is 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.