Learn More
TARGETMOL CHEMICALS INC BI-749327 1MG
SDP
Catalog No. 502880778
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. BI-749327 is a high selectivity and orally bioavailable antagonist of TRPC6 (IC50s 13 nM, 19 nM and 15 nM for mouse, human and guinea pig TRPC6). Purity 99.68%
Catalog No. 50-288-0778 Supplier TARGETMOL CHEMICALS INC Supplier No. T105371MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. BI-749327 is a high selectivity and orally bioavailable antagonist of TRPC6 (IC50s 13 nM, 19 nM and 15 nM for mouse, human and guinea pig TRPC6). Purity 99.68%