Learn More
Medchemexpress LLC BI-749327 | 2361241-23-6 | 99.9% | 442.43 | 1 MG
SDP
Catalog No. 5000439496
Encompass_Preferred
BI-749327 is a potent, highly selective, and orally bioavailable TRPC6 antagonist. It demonstrates significant selectivity, being 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7. In vitro, it suppresses NFAT activation and blocks signaling of prohypertrophic genes in isolated myocytes.
  • Potent and selective TRPC6 antagonist
  • Orally bioavailable
  • Suppresses NFAT activation in HEK293T cells
  • Blocks prohypertrophic gene expression in isolated myocytes
  • Improves left heart function and reduces volume/mass ratio in vivo
  • Reduces renal fibrosis in vivo
  • Long terminal half-life in mice
Catalog No. 50-004-39496 Supplier Medchemexpress LLC Supplier No. HY1119251MG
BI-749327 is a potent, highly selective, and orally bioavailable TRPC6 antagonist. It demonstrates significant selectivity, being 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7. In vitro, it suppresses NFAT activation and blocks signaling of prohypertrophic genes in isolated myocytes.
  • Potent and selective TRPC6 antagonist
  • Orally bioavailable
  • Suppresses NFAT activation in HEK293T cells
  • Blocks prohypertrophic gene expression in isolated myocytes
  • Improves left heart function and reduces volume/mass ratio in vivo
  • Reduces renal fibrosis in vivo
  • Long terminal half-life in mice