Learn More
Medchemexpress LLC BI-749327 | 2361241-23-6 | 99.9% | 442.43 | 100 MG
SDP
Catalog No. 5000439460
Encompass_Preferred
BI-749327 is a potent, highly selective, and orally bioavailable TRPC6 antagonist, with IC50s of 13 nM, 19 nM, and 15 nM for mouse, human, and guinea pig TRPC6, respectively. It is 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7.
  • Potent, highly selective, and orally bioavailable TRPC6 antagonist.
  • Suppresses NFAT activation in HEK293T cells.
  • Blocks associated signaling and expression of prohypertrophic genes in isolated myocytes.
  • Improves left heart function.
  • Reduces renal fibrosis.
Catalog No. 50-004-39460 Supplier Medchemexpress LLC Supplier No. HY111925100MG
BI-749327 is a potent, highly selective, and orally bioavailable TRPC6 antagonist, with IC50s of 13 nM, 19 nM, and 15 nM for mouse, human, and guinea pig TRPC6, respectively. It is 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7.
  • Potent, highly selective, and orally bioavailable TRPC6 antagonist.
  • Suppresses NFAT activation in HEK293T cells.
  • Blocks associated signaling and expression of prohypertrophic genes in isolated myocytes.
  • Improves left heart function.
  • Reduces renal fibrosis.