Learn More
Medchemexpress LLC BI-765063 10mg | 10mg
SDP
Catalog No. 5000325598
Encompass_Preferred
BI-765063 (OSE-172) is a humanized antibody expressed in CHO that targets SIRPa/CD172a BI-765063 contains an IgG4-type heavy chain and a hu -type light chain with a predicted molecular weight (MW) of 146 3 kDa The isotype control for BI-765063 can be referenced as Human IgG4 kappa Isotype Control (HY-P99003)
Catalog No. 50-003-25598 Supplier Medchemexpress LLC Supplier No. HYP99055510MG
BI-765063 (OSE-172) is a humanized antibody expressed in CHO that targets SIRPa/CD172a BI-765063 contains an IgG4-type heavy chain and a hu -type light chain with a predicted molecular weight (MW) of 146 3 kDa The isotype control for BI-765063 can be referenced as Human IgG4 kappa Isotype Control (HY-P99003)