Learn More
Medchemexpress LLC HY-10366 5mg Medchemexpress, BI-78D3 CAS:883065-90-5 Purity:>98%
SDP
Catalog No. 501968151
Encompass_Preferred
Medchemexpress, HY-10366 5mg BI-78D3 CAS:883065-90-5 BI-78D3 functions as a substrate competitive inhibitor of JNK, inhibit the JNK kinase activity (IC50=280 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8151 Supplier Medchemexpress LLC Supplier No. HY103665MG
Medchemexpress, HY-10366 5mg BI-78D3 CAS:883065-90-5 BI-78D3 functions as a substrate competitive inhibitor of JNK, inhibit the JNK kinase activity (IC50=280 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.