Learn More
Selleck Chemical LLC BI-78D3
SDP
Catalog No. 5000001747
Encompass_Preferred
BI-78D3 is a competitive JNK inhibitor with IC50 of 280 nM that displays 100 fold selectivity over p38 and no activity at mTOR and PI-3K
Catalog No. 50-000-01747 Supplier Selleck Chemical LLC Supplier No. S820125MG
BI-78D3 is a competitive JNK inhibitor with IC50 of 280 nM that displays 100 fold selectivity over p38 and no activity at mTOR and PI-3K